Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid3a
Entrez Gene ID
314616
Gene Name
AT rich interactive domain 3A (Bright like)
Gene Aliases
-
Chromosome Location
Chr.7: 12575989 - 12602415 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765169.1 XP_008763391.1 1 - 2 77 59

View Full Product Details