Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chmp6
Entrez Gene ID
287873
Gene Name
charged multivesicular body protein 6
Gene Aliases
RGD1565325
Chromosome Location
Chr.10: 109073678 - 109079348 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105856.1 NP_001099326.1 6 - 7 537 61
GenBank mRNA BC070963.1 - 4 - 5 329 61

View Full Product Details