The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008283.1 | NP_001008284.1 | 4 - 5 | 587 | 102 |
XM_006247798.1 | XP_006247860.1 | 4 - 5 | 573 | 102 | |
GenBank mRNA | BC086594.1 | - | 4 - 5 | 587 | 102 |