Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bst1
Entrez Gene ID
81506
Gene Name
bone marrow stromal cell antigen 1
Gene Aliases
-
Chromosome Location
Chr.14: 71798218 - 71814540 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030848.1 NP_110475.1 2 - 3 339 108
GenBank mRNA D49955.1 - 2 - 3 339 108

View Full Product Details