Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Poln
Entrez Gene ID
102555079
Gene Name
polymerase (DNA directed) nu
Gene Aliases
-
Chromosome Location
Chr.14: 81837835 - 82037999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006251412.1 XP_006251474.1 22 - 23 2892 90

View Full Product Details