Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Poln
Entrez Gene ID
102555079
Gene Name
polymerase (DNA directed) nu
Gene Aliases
-
Chromosome Location
Chr.14: 81837835 - 82037999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006251412.1 XP_006251474.1 10 - 11 1900 102
XM_006251414.2 XP_006251476.1 10 - 11 1900 102
XM_008770382.1 XP_008768604.1 10 - 11 1900 102
XM_008770383.1 XP_008768605.1 10 - 11 1459 102
XM_008770384.1 XP_008768606.1 10 - 11 1900 102

View Full Product Details