Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ldb3
Entrez Gene ID
498587
Gene Name
LIM domain binding 3
Gene Aliases
RGD1564875
Chromosome Location
Chr.16: 10878355 - 10941464 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110490.2 NP_001103960.1 2 - 3 184 130
NM_001277158.1 NP_001264087.1 2 - 3 184 130
NM_001277159.1 NP_001264088.1 2 - 3 174 130
NM_001277165.1 NP_001264094.1 2 - 3 174 130
NM_001277166.1 NP_001264095.1 2 - 3 174 130
XM_006252637.2 XP_006252699.1 2 - 3 230 130
XM_006252640.2 XP_006252702.1 2 - 3 230 130
XM_006252641.2 XP_006252703.1 2 - 3 230 130
XM_006252644.2 XP_006252706.1 2 - 3 230 130
XM_006252645.2 XP_006252707.1 2 - 3 230 130
XM_006252647.1 XP_006252709.1 2 - 3 230 130
GenBank mRNA BC083721.1 - 1 - 2 122 130
FN804937.1 - 2 - 3 142 130
FQ215617.1 - 2 - 3 170 130
FQ216341.1 - 2 - 3 180 130
FQ216348.1 - 2 - 3 180 130
FQ217970.1 - 2 - 3 180 130
FQ223559.1 - 2 - 3 180 130

View Full Product Details