Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eaf1
Entrez Gene ID
306261
Gene Name
ELL associated factor 1
Gene Aliases
-
Chromosome Location
Chr.16: 7588217 - 7602966 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107293.1 NP_001100763.1 4 - 5 1426 69

View Full Product Details