Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcp1a
Entrez Gene ID
361109
Gene Name
decapping mRNA 1A
Gene Aliases
RGD1561298
Chromosome Location
Chr.16: 6541865 - 6583856 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191831.1 NP_001178760.1 1 - 2 196 98

View Full Product Details