Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf7
Entrez Gene ID
364510
Gene Name
PHD finger protein 7
Gene Aliases
-
Chromosome Location
Chr.16: 7323527 - 7336230 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012211.1 NP_001012211.1 2 - 3 193 91
XM_006252632.2 XP_006252694.1 1 - 2 113 91
GenBank mRNA BC079288.1 - 2 - 3 193 91

View Full Product Details