Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wasf3
Entrez Gene ID
682937
Gene Name
WAS protein family, member 3
Gene Aliases
-
Chromosome Location
Chr.12: 10298688 - 10368620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769055.1 XP_008767277.1 8 - 9 2187 83

View Full Product Details