Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc29a4
Entrez Gene ID
288499
Gene Name
solute carrier family 29 (equilibrative nucleoside transporter), member 4
Gene Aliases
-
Chromosome Location
Chr.12: 13914114 - 13924511 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105911.1 NP_001099381.1 8 - 9 1636 66

View Full Product Details