Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Get4
Entrez Gene ID
288518
Gene Name
golgi to ER traffic protein 4 homolog (S. cerevisiae)
Gene Aliases
Cee, RGD1311660
Chromosome Location
Chr.12: 17469112 - 17488396 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163320.1 NP_001156792.1 2 - 3 350 92

View Full Product Details