Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fads2
Entrez Gene ID
83512
Gene Name
fatty acid desaturase 2
Gene Aliases
Fadsd6
Chromosome Location
Chr.1: 225906582 - 226152568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031344.2 NP_112634.1 5 - 6 904 105
XM_008760244.1 XP_008758466.1 5 - 6 2413 105
GenBank mRNA AB021980.1 - 5 - 6 834 105
BC081776.1 - 5 - 6 904 105
FQ210893.1 - 5 - 6 833 105
HQ909027.1 - 5 - 6 738 105

View Full Product Details