Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmp21
Entrez Gene ID
293573
Gene Name
matrix metallopeptidase 21
Gene Aliases
-
Chromosome Location
Chr.1: 205745119 - 205750786 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106308.1 NP_001099778.1 3 - 4 851 65

View Full Product Details