The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009706.1 | NP_001009706.1 | 4 - 5 | 541 | 66 |
XM_008759911.1 | XP_008758133.1 | 4 - 5 | 568 | 66 | |
XM_008759912.1 | XP_008758134.1 | 4 - 5 | 1926 | 66 | |
GenBank mRNA | BC088448.1 | - | 4 - 5 | 541 | 66 |