Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fxn
Entrez Gene ID
499335
Gene Name
frataxin
Gene Aliases
RGD1565754
Chromosome Location
Chr.1: 242123975 - 242153168 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760308.1 XP_008758530.1 1 - 1 168 101

View Full Product Details