The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108507.2 | NP_001101977.1 | 2 - 3 | 1544 | 60 |
GenBank mRNA | BC162015.1 | - | 2 - 3 | 1544 | 60 |