Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnh1
Entrez Gene ID
100360501
Gene Name
ribonuclease/angiogenin inhibitor 1
Gene Aliases
Rnh
Chromosome Location
Chr.1: 214151337 - 214163807 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270762.1 NP_001257691.1 3 - 4 376 114
NM_001270763.1 NP_001257692.1 3 - 4 382 114
NM_139105.3 NP_620805.2 3 - 4 391 114
XM_008759970.1 XP_008758192.1 3 - 4 940 114
GenBank mRNA AB296102.1 - 2 - 3 258 114
BC070501.1 - 3 - 4 298 114
FQ233708.1 - 3 - 4 316 114
FQ234889.1 - 3 - 4 315 114
X62528.1 - 3 - 4 376 114

View Full Product Details