Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnh1
Entrez Gene ID
100360501
Gene Name
ribonuclease/angiogenin inhibitor 1
Gene Aliases
Rnh
Chromosome Location
Chr.1: 214151337 - 214163807 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270762.1 NP_001257691.1 9 - 10 1404 97
NM_001270763.1 NP_001257692.1 9 - 10 1410 97
NM_139105.3 NP_620805.2 9 - 10 1419 97
XM_008759970.1 XP_008758192.1 9 - 10 1968 97
GenBank mRNA AB296102.1 - 8 - 9 1286 97
BC070501.1 - 9 - 10 1326 97
FQ233708.1 - 9 - 10 1344 97
FQ234889.1 - 9 - 10 1346 97
X62528.1 - 9 - 10 1404 97

View Full Product Details