The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106372.2 | NP_001099842.1 | 3 - 4 | 649 | 79 |
GenBank mRNA | BC158801.1 | - | 3 - 4 | 649 | 79 |