Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d10c
Entrez Gene ID
361697
Gene Name
TBC1 domain family, member 10C
Gene Aliases
RGD1311490
Chromosome Location
Chr.1: 219431094 - 219438839 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081980.1 NP_001075449.1 9 - 10 1036 67
XM_006230840.2 XP_006230902.1 9 - 10 1089 67
GenBank mRNA BC160813.1 - 9 - 10 1047 67
CO564143.1 - - 562 67
FQ226277.1 - 9 - 10 1047 67

View Full Product Details