The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009603.1 | NP_001009603.1 | 3 - 4 | 633 | 73 |
XM_006230733.2 | XP_006230795.1 | 3 - 4 | 642 | 73 | |
XM_006230734.2 | XP_006230796.1 | 3 - 4 | 695 | 73 | |
XM_008760098.1 | XP_008758320.1 | 3 - 4 | 803 | 73 | |
XM_008760099.1 | XP_008758321.1 | 3 - 4 | 695 | 73 | |
GenBank mRNA | BC088190.1 | - | 3 - 4 | 633 | 73 |