Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ahnak
Entrez Gene ID
191572
Gene Name
AHNAK nucleoprotein
Gene Aliases
AHNAK 1, RGD1308572, UV118
Chromosome Location
Chr.1: 225184901 - 225215015 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191951.1 NP_001178880.1 6 - 6 4429 91
XM_008760214.1 XP_008758436.1 5 - 5 4830 91
GenBank mRNA DQ203292.1 - 1 - 1 91
DQ203294.1 - 1 - 1 91

View Full Product Details