The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108481.1 | NP_001101951.1 | 3 - 4 | 357 | 66 |
GenBank mRNA | FQ211166.1 | - | 3 - 4 | 373 | 66 |