Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2g1
Entrez Gene ID
25251
Gene Name
cytochrome P450, family 2, subfamily g, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.1: 83626639 - 83637780 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012787.1 NP_036919.1 2 - 3 342 118
GenBank mRNA M33296.2 - 2 - 3 340 118

View Full Product Details