Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp5sl
Entrez Gene ID
361520
Gene Name
ATP5S-like
Gene Aliases
-
Chromosome Location
Chr.1: 82413223 - 82419157 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009705.1 NP_001009705.1 4 - 5 421 74
XM_006228449.2 XP_006228511.1 4 - 5 548 74
GenBank mRNA BC088283.1 - 4 - 5 421 74

View Full Product Details