The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009705.1 | NP_001009705.1 | 5 - 6 | 590 | 82 |
XM_006228449.2 | XP_006228511.1 | 5 - 6 | 717 | 82 | |
GenBank mRNA | BC088283.1 | - | 5 - 6 | 590 | 82 |