Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc9
Entrez Gene ID
684934
Gene Name
coiled-coil domain containing 9
Gene Aliases
-
Chromosome Location
Chr.1: 78212933 - 78226400 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759030.1 XP_008757252.1 12 - 13 1700 150

View Full Product Details