Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adck4
Entrez Gene ID
308453
Gene Name
aarF domain containing kinase 4
Gene Aliases
-
Chromosome Location
Chr.1: 84044551 - 84067066 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012065.1 NP_001012065.1 13 - 14 1279 77
GenBank mRNA BC079227.1 - 13 - 14 1279 77

View Full Product Details