The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133600.1 | NP_598284.1 | 4 - 5 | 533 | 101 |
GenBank mRNA | AF268030.1 | - | 4 - 5 | 533 | 101 |
AY539951.1 | - | 3 - 4 | 365 | 101 | |
BC078745.1 | - | 4 - 5 | 495 | 101 |