The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109511.1 | NP_001102981.1 | 1 - 2 | 117 | 102 |
GenBank mRNA | BC166898.1 | - | 1 - 2 | 114 | 102 |