Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map9
Entrez Gene ID
310544
Gene Name
microtubule-associated protein 9
Gene Aliases
Mtap9, RGD1560368
Chromosome Location
Chr.2: 181331438 - 181368087 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135716.1 NP_001129188.1 12 - 13 1885 79
XM_006232515.2 XP_006232577.1 12 - 13 1947 79
XM_006232516.2 XP_006232578.1 11 - 12 1799 79

View Full Product Details