Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map9
Entrez Gene ID
310544
Gene Name
microtubule-associated protein 9
Gene Aliases
Mtap9, RGD1560368
Chromosome Location
Chr.2: 181331438 - 181368087 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232515.2 XP_006232577.1 1 - 2 193 143

View Full Product Details