Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpini2
Entrez Gene ID
171149
Gene Name
serpin peptidase inhibitor, clade I (pancpin), member 2
Gene Aliases
-
Chromosome Location
Chr.2: 173640385 - 173670794 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134409.1 NP_001127881.1 7 - 8 1147 136
XM_008761077.1 XP_008759299.1 8 - 9 1320 136
GenBank mRNA X99773.1 - 7 - 8 1162 136
Z30585.1 - 5 - 6 515 136

View Full Product Details