The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033895.1 | NP_001029067.1 | 1 - 2 | 157 | 90 |
XM_008761146.1 | XP_008759368.1 | 1 - 2 | 271 | 90 | |
GenBank mRNA | BC128708.1 | - | 1 - 2 | 282 | 90 |
BC168242.1 | - | 1 - 2 | 270 | 90 | |
DQ185515.1 | - | 1 - 2 | 157 | 90 |