Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crtc2
Entrez Gene ID
310615
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
RGD1308903, Torc2
Chromosome Location
Chr.2: 189615940 - 189626075 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033895.1 NP_001029067.1 9 - 10 759 76
XM_008761146.1 XP_008759368.1 8 - 9 804 76
GenBank mRNA BC128708.1 - 9 - 10 884 76
BC168242.1 - 9 - 10 872 76
DQ185515.1 - 9 - 10 759 76

View Full Product Details