The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024797.1 | NP_001019968.1 | 19 - 20 | 2778 | 92 |
XM_006232735.2 | XP_006232797.1 | 19 - 20 | 2769 | 92 | |
XM_006232736.2 | XP_006232798.1 | 19 - 20 | 2754 | 92 | |
XM_006232737.2 | XP_006232799.1 | 18 - 19 | 2733 | 92 | |
XM_006232738.2 | XP_006232800.1 | 18 - 19 | 2730 | 92 | |
XM_006232739.2 | XP_006232801.1 | 19 - 20 | 2709 | 92 | |
XM_006232740.2 | XP_006232802.1 | 18 - 19 | 2565 | 92 | |
XM_006232741.2 | XP_006232803.1 | 19 - 20 | 2757 | 92 | |
XM_008761201.1 | XP_008759423.1 | 20 - 21 | 2783 | 92 | |
XM_008761202.1 | XP_008759424.1 | 18 - 19 | 2218 | 92 | |
GenBank mRNA | AB195689.1 | - | 18 - 19 | 2612 | 92 |
AY724781.1 | - | 19 - 20 | 2778 | 92 | |
AY724782.1 | - | 19 - 20 | 2778 | 92 |