The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033896.1 | NP_001029068.1 | 3 - 4 | 1179 | 60 |
XM_006232637.2 | XP_006232699.1 | 3 - 4 | 1233 | 60 | |
XM_008761149.1 | XP_008759371.1 | 2 - 3 | 1698 | 60 | |
GenBank mRNA | BC101913.1 | - | 3 - 4 | 1179 | 60 |