Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubn2
Entrez Gene ID
312248
Gene Name
ubinuclein 2
Gene Aliases
RGD1310722
Chromosome Location
Chr.4: 66164762 - 66254297 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134553.1 NP_001128025.1 6 - 7 1598 140
XM_006236308.2 XP_006236370.1 6 - 7 2681 140

View Full Product Details