Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1galt1
Entrez Gene ID
65044
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1
Gene Aliases
-
Chromosome Location
Chr.4: 33890089 - 33943219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022950.1 NP_075239.1 2 - 3 377 102
XM_006236091.2 XP_006236153.1 3 - 4 398 102
XM_008762725.1 XP_008760947.1 3 - 4 347 102
GenBank mRNA AF157963.1 - 2 - 3 377 102

View Full Product Details