The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183328.2 | NP_899157.1 | 2 - 3 | 82 | 65 |
XM_008762822.1 | XP_008761044.1 | 3 - 4 | 328 | 65 | |
XM_008762823.1 | XP_008761045.1 | 3 - 4 | 264 | 65 | |
XM_008762824.1 | XP_008761046.1 | 3 - 4 | 208 | 65 | |
XM_008762825.1 | XP_008761047.1 | 3 - 4 | 196 | 65 | |
XM_008762826.1 | XP_008761048.1 | 3 - 4 | 147 | 65 | |
XM_008762827.1 | XP_008761049.1 | 3 - 4 | 328 | 65 | |
XM_008762828.1 | XP_008761050.1 | 3 - 4 | 203 | 65 | |
XM_008762829.1 | XP_008761051.1 | 2 - 3 | 127 | 65 | |
GenBank mRNA | M94557.1 | - | 2 - 3 | 136 | 65 |