Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlx6
Entrez Gene ID
500023
Gene Name
distal-less homeobox 6
Gene Aliases
RGD1561539
Chromosome Location
Chr.4: 32373027 - 32377388 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762741.1 XP_008760963.1 1 - 2 694 96

View Full Product Details