Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gdpd1
Entrez Gene ID
303407
Gene Name
glycerophosphodiester phosphodiesterase domain containing 1
Gene Aliases
RGD1311813
Chromosome Location
Chr.10: 74334972 - 74376332 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044238.1 NP_001037703.1 6 - 7 767 97
GenBank mRNA BC105620.1 - 6 - 7 767 97

View Full Product Details