Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlgn2
Entrez Gene ID
117096
Gene Name
neuroligin 2
Gene Aliases
-
Chromosome Location
Chr.10: 56431586 - 56444847 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053992.1 NP_446444.1 2 - 3 1012 93
GenBank mRNA U41662.1 - 2 - 3 1012 93

View Full Product Details