The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109035.1 | NP_001102505.1 | 7 - 8 | 1145 | 92 |
XM_006246786.2 | XP_006246848.1 | 8 - 9 | 1734 | 92 | |
XM_006246789.2 | XP_006246851.1 | 7 - 8 | 1582 | 92 | |
XM_008767857.1 | XP_008766079.1 | 8 - 9 | 1735 | 92 |