The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107002.1 | NP_001100472.1 | 9 - 10 | 1407 | 73 |
XM_006246271.2 | XP_006246333.1 | 9 - 10 | 1420 | 73 |