Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc57
Entrez Gene ID
688324
Gene Name
coiled-coil domain containing 57
Gene Aliases
-
Chromosome Location
Chr.10: 110006274 - 110100125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768498.1 XP_008766720.1 10 - 11 1742 86

View Full Product Details