The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019201.3 | NP_062074.2 | 8 - 9 | 1373 | 122 |
XM_006251225.2 | XP_006251287.1 | 7 - 8 | 1208 | 122 | |
XM_006251226.2 | XP_006251288.1 | 7 - 8 | 1208 | 122 | |
XM_006251227.2 | XP_006251289.1 | 7 - 8 | 1207 | 122 | |
XM_006251228.2 | XP_006251290.1 | 7 - 8 | 1208 | 122 | |
XM_006251229.2 | XP_006251291.1 | 8 - 9 | 1399 | 122 | |
GenBank mRNA | AF067795.2 | - | 8 - 9 | 1378 | 122 |
BC078778.1 | - | 8 - 9 | 1384 | 122 |