The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191606.1 | NP_001178535.1 | 3 - 4 | 4283 | 86 |
XM_008760920.1 | XP_008759142.1 | 4 - 5 | 5231 | 86 | |
XM_008760921.1 | XP_008759143.1 | 4 - 5 | 4903 | 86 | |
XM_008760922.1 | XP_008759144.1 | 4 - 5 | 4825 | 86 |